missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Rabbit IgG
This Antibody Labeling Kit conjugates CoraLite™ 750 into Rabbit IgG antibodies.
£75.00 - £710.00
Specifications
Content And Storage | 1 Year at -20°C or 6 Months at +4°C, Avoid Light |
---|---|
Excitation/Emission | 755 nm/780 nm |
Labeling Target | IgG |
Species | Rabbit |
Product Code | Brand | Kit Contents | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Kit Contents | Price | Quantity & Availability | |||||
17731519
|
Proteintech™
KFA004-10 |
10 μL CoraLite™ Plus 750 FlexLinker for Rabbit IgG, 20 μL FlexQuencher for Rabbit IgG, 100 μL FlexBuffer, Manual, Protocol Magnet |
£75.00
10 reactions |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
17751519
|
Proteintech™
KFA004-50 |
50 μL CoraLite™ Plus 750 FlexLinker for Rabbit IgG, 100 μL FlexQuencher for Rabbit IgG, 500 μL FlexBuffer, Manual, Protocol Magnet |
£335.00
50 reactions |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
17741519
|
Proteintech™
KFA004-200 |
4 x 50 μL CoraLite™ Plus 750 FlexLinker for Rabbit IgG, 4 x 100 μL FlexQuencher for Rabbit IgG, 4 x 500 μL FlexBuffer, Manual, Protocol Magnet |
£710.00
200 reactions |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 750 in any buffer condition to rabbit IgG primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your rabbit IgG antibody in <10 min without any additional equipment
Specifications
1 Year at -20°C or 6 Months at +4°C, Avoid Light | |
755 nm/780 nm | |
IgG | |
Rabbit |