Learn More
Fisherbrand™ Test Tube Holder for Ultrasonic Units
Use this Fisherbrand™ Test Tube Holder with Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. Easily support and handle test tubes within the ultrasonic cleaner.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.26000kg
Description
- Multiple holes to support 12 test tubes
- Dual handles easily secures holder in the ultrasonic cleaner
- Stainless steel
- Light and easy to maneuver
Specifications
Test Tube Holder | |
FB11211 Ultrasonic Cleaning Unit | |
Stainless Steel |