missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Comb One Well 70mm x 1.5mm Preparative for Vari-Gel™ Tank
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.80000kg
Description
• Three different Vari-gel tanks, 150mm, 100mm and 70mm are available accommodating up to four different gel tray sizes• UV transparent gel trays allow for placement directly onto a UV transilluminator for gel viewing once stained
• Asymmetric electrode offset - prevents accidental polarity reversal ensuring no possibility of sample loss into the buffer
• aluminum casting gates - ensure easy casting of the gel inside or outside of the tank. They slide into pre-formed grooves at the end of the gel tray providing a secure seal against gel leakage
• Different gel sizes can be interchanged in one tank, combining superb resolution with ease of use.
Specifications
Vari-Gel™ Tank | |
Comb Preparative | |
1 | |
70 x 1.5 mm |