Learn More
Fisher BioReagents™ EZ-Run™ Protein Gel Staining Solution
Ready-to-use solution for highly sensitive staining of proteins separated in polyacrylamide gels
Brand: Fisher BioReagents™ BP3620-4
Code : 32
Additional Details : CAS Number : 67-63-0 Weight : 4.05000kg
Description
EZ-Run Protein Gel Staining Solution contains Coomassie™ Brilliant Blue G-250. Due to the property of the dye to form colloidal particles, proteins are preferentially stained without significant background staining of the gel matrix.
- Detection is linear over two orders of magnitude with as little as 5ng protein detected
- Sensitivity: 5ng BSA in 10% SDS-PAGE, 1.0mm thick gel, well width 3mm
- Does not contain toxic methanol or acetic acid
- Provides rapid destaining with simple water washes
- Can be reused twice without significant loss of sensitivity
- Tested for staining sensitivity in minigels using purified proteins and Protein Molecular Weight Standards
Specifications
Dark Blue | |
Liquid | |
Highly sensitive staining of proteins separated in Polyacrylamide gels |
Stain | |
Coomassie™ Brilliant Blue G-250 | |
4 L |