Learn More
Thermo Scientific™ Finntip™ Biocon Pipette Tips
Perform PCR and other purity demanding applications with these biocon pipette tips, certified free from endotoxin, DNA, DNase and RNase.
Brand:  Thermo Scientific™ 94053100
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10115582
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Features:
  • Individually packed
  • Three models in volume range 0.2-1000μL
  • Presterilized with gamma radiation
  • Includes certificate of quality
Recommended for: PCR applications, molecular biology
Specifications
Specifications
Finntip BioCon 10, Non-Filtered, Sterile,
Non-Filtered
Finntip BioCon 10
Thermo Scientific FinnTip BioCon 10
10 μL
Sterile
ISO 9001, ISO 14001, ISO 13485
FinnTip Biocon