Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, United States
Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Brand: Gibco™ A3840101
52.96 GBP valid until 2024-12-31
Use promo code "21878" to get your promotional price.
Additional Details : Weight : 0.90000kg
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Value Plus (Performance)
- Origin: United States.
- Endotoxin level: ≤10 EU/mL
- Hemoglobin level: ≤25 mg/dL
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Fetal | |
United States | |
One Shot | |
50 mL | |
Sterile | |
≤10 EU/mL | |
Qualified | |
Frozen |
Fetal Bovine Serum, heat inactivated, qualified, One Shot™, United States | |
Bovine | |
Liquid | |
Fetal Bovine Serum | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
≤25 mg/dL | |
One Shot™ |