missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM114A1 Mouse anti-Human, Unconjugated, Clone: 1B11D6, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | FAM114A1 |
---|---|
Clone | 1B11D6 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17561436
|
Proteintech
67926-1-IG-20UL |
20 μL |
£130.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17551436
|
Proteintech
67926-1-IG-150UL |
150 μL |
£339.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FAM114A1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human FAM114A1. This antibody recognizes human, rat, mouse antigen. The FAM114A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
FAM114A1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse, Rat | |
IgG2a |
1B11D6 | |
Monoclonal | |
Mouse | |
92689 | |
Primary |