Learn More
EXTL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558086
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ALPPRPRPGA SQGWPRWLDA ELLQSFSQPG ELPEDAVSPP QAPHGGSCNW ESCFDTSKCR GDGLKVFVYP AVGTISETHR RILASIEGS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.
This gene is a member of the multiple exostoses (EXT) family of glycosyltransferases, which function in the chain polymerization of heparan sulfate and heparin. The encoded protein harbors alpha 1,4- N-acetylglucosaminyltransferase activity, and is involved in chain elongation of heparan sulfate and possibly heparin.Specifications
EXTL1 | |
Polyclonal | |
Unconjugated | |
Extl1 | |
alpha-N-acetylglucosaminyltransferase II; D430033M16Rik; EC 2.4.1.224; exostoses (multiple)-like 1; exostoses-like 1; exostosin like glycosyltransferase 1; exostosin-L; Exostosin-like 1; Exostosin-like 1 (Exostosin-L) (Multiple exostosis-like protein); exostosin-like glycosyltransferase 1; EXTL; Extl1; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Glucuronosyl-N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase; glucuronyl-N-acetylglucosaminylproteoglycan alpha-1,4-N- acetylglucosaminyltransferase; MGC70794; multiple exostoses-like 1; multiple exostosis-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92935 | |
Extl1 | |
Recombinant protein corresponding to Human EXTL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only