Learn More
EXOC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565195
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VENLKNIFSVP EIVRETQDLI EQGALLQAHR KLMDLECSRD GLMYEQYRMD SGNTRDMTLI HGYFGSTQGL SDELAKQLWM V Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.Specifications
EXOC3 | |
Polyclonal | |
Unconjugated | |
EXOC3 | |
2810050O03Rik; E430013E20Rik; EXOC3; EXOC3L; EXOC3L1; Exocyst complex component 3; exocyst complex component 3 like 1; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Exocyst complex component Sec6; I79_024581; KIAA0895L; KIAA0895-like ortholog; LIM/homeobox protein Lhx2 pseudogene; protein Jiangli; RGD1311132; rSec6; Sec 6 homolog; Sec6; Sec6 protein; SEC6L1; SEC6-like 1; Sec6p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60645 | |
EXOC3 | |
Recombinant Human EXOC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only