Learn More
Thermo Scientific™ EVOS™ Power Cord, Type L (Italy)
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type L power plugs.
Brand: Thermo Scientific™ AMEP4648
Code : NEW
Additional Details : Weight : 0.50000kg
Description
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type L power plugs. The Type L plug is used in Italy, Chile, Uruguay, and parts of North Africa.
Specifications
Power Cord | |
Type L (Plug), Type L (Power Plug) | |
1 Ea. |
EVOS | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |