Learn More
EVOS™ Olympus 60X Oil Objective, apochromat, coverslip-corrected
For demanding applications, especially capturing color images in white light
Brand:  EVOS™ AMEP4694
Product Code. 11907315
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Can be used to view images with different magnification
  • Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Objective Lens
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Plan Apochromat
1 each
Olympus
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
60X