missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Ethyl mandelate, 97%, Thermo Scientific Chemicals

£31.20 - £85.70

Chemical Identifiers

CAS 774-40-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00004494
InChI Key SAXHIDRUJXPDOD-UHFFFAOYSA-N
Synonym ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
PubChem CID 13050
ChEBI CHEBI:38750
IUPAC Name ethyl 2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11408007
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17412.14
25 g
£31.20
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11418007
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17412.22
100 g
£85.70
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

774-40-3
180.203
SAXHIDRUJXPDOD-UHFFFAOYSA-N
13050
ethyl 2-hydroxy-2-phenylacetate
C10H12O3
MFCD00004494
ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
CHEBI:38750
CCOC(=O)C(C1=CC=CC=C1)O

Specifications

774-40-3
253°C to 255°C
C10H12O3
ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
CCOC(=O)C(C1=CC=CC=C1)O
180.203
CHEBI:38750
97%
1.115
>110°C (230°F)
MFCD00004494
SAXHIDRUJXPDOD-UHFFFAOYSA-N
ethyl 2-hydroxy-2-phenylacetate
13050
180.2
(+/-)-Ethyl mandelate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

missing translation for 'einecsNumber' : 212-263-0

missing translation for 'rtecsNumber' : OO7397000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only