missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, BAKER ULTRA RESI-ANALYZED, J.T. Baker™
205.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.
- Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
- Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
- Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
- Performance is guaranteed through the expiration period stated on the label
- Suitable for exacting EPA sample cleanup protocols
Specifications
Melting Point | -83.57°C |
Color | Colorless |
Linear Formula | CH3COOC2H5 |
Solubility Information | 79g/L in water |
Formula Weight | 88.11g/mol |
Percent Purity | ≥99.6% |
Grade | For Organic Residue Analysis |
Density | 0.9g/cm3 |
Boiling Point | 76°C to 78°C |
Quantity | 1 L |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'