missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, BAKER, J.T. Baker™

Product Code. 10382681
2.5 L, Glass
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in Histology applications.

A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).

Specifications

Melting Point -83.57°C
CAS 141-78-6
Color Colorless
Appearance Passes test
Density 0.9g/cm3 at 20°C
Boiling Point 76°C to 78°C
Quantity 2.5 L
Explosion Limit 2.1 - 11.5 Vol %
Flash Point −1°C
Ignition Point 460°C
Odor Fruit-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 0.003%
Molecular Formula C4H8O2
Viscosity 0.44 mPas at 20°C
Water Max. 0.1%
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
Solubility Information 79g/L in water
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES CCOC(=O)C
IUPAC Name ethyl acetate
Molecular Weight (g/mol) 88.106
PubChem CID 8857
ChEBI CHEBI:27750
Percent Purity ≥99%
Grade Manufactured in US FDA / ISO 9000 facility, cGMP, IPEC member
Chemical Name or Material Ethyl Acetate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl Acetate, BAKER, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.