missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, BAKER ANALYZED, J.T. Baker™
168.15 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
2.5 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
Description
- Exceeds the requirements set forth by the American Chemical Society (ACS)
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Melting Point | -83.57°C |
Color | Colorless |
Linear Formula | CH3COOC2H5 |
Solubility Information | 79g/L (in water) |
Formula Weight | 88.11g/mol |
Percent Purity | ≥99.5% |
Grade | Exceeds ACS specifications |
Density | 0.9g/cm3 at 20°C |
Boiling Point | 76°C to 78°C |
Quantity | 2.5 L |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'