Learn More
Ethyl 2-morpholinobenzoate, 97%, Thermo Scientific™
£27.60 - £380.00
Chemical Identifiers
CAS | 192817-79-1 |
---|---|
Molecular Formula | C13H17NO3 |
Molecular Weight (g/mol) | 235.283 |
MDL Number | MFCD06204497 |
InChI Key | UIVDSGDHUXOYDW-UHFFFAOYSA-N |
Synonym | ethyl 2-morpholinobenzoate, ethyl 2-morpholin-4-yl benzoate, 2-morpholin-4-yl-benzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-ylbenzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-, ethyl ester |
PubChem CID | 7148401 |
IUPAC Name | ethyl 2-morpholin-4-ylbenzoate |
SMILES | CCOC(=O)C1=CC=CC=C1N2CCOCC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10537873
|
Thermo Scientific Maybridge
CC24323CB |
250 mg |
£27.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10127762
|
Thermo Scientific Maybridge
CC24323DA |
1 g |
£75.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10012374
|
Thermo Scientific Maybridge
CC24323EA |
10 g |
£380.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
192817-79-1 | |
235.283 | |
UIVDSGDHUXOYDW-UHFFFAOYSA-N | |
7148401 | |
CCOC(=O)C1=CC=CC=C1N2CCOCC2 |
C13H17NO3 | |
MFCD06204497 | |
ethyl 2-morpholinobenzoate, ethyl 2-morpholin-4-yl benzoate, 2-morpholin-4-yl-benzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-ylbenzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-, ethyl ester | |
ethyl 2-morpholin-4-ylbenzoate |
Specifications
192817-79-1 | |
100.0 | |
Amber glass bottle | |
MFCD06204497 | |
UIVDSGDHUXOYDW-UHFFFAOYSA-N | |
ethyl 2-morpholin-4-ylbenzoate | |
7148401 | |
97% |
96.5 | |
97% | |
C13H17NO3 | |
ethyl 2-morpholinobenzoate, ethyl 2-morpholin-4-yl benzoate, 2-morpholin-4-yl-benzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-ylbenzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-, ethyl ester | |
CCOC(=O)C1=CC=CC=C1N2CCOCC2 | |
235.283 | |
235.28 | |
Ethyl 2-morpholinobenzoate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.