Learn More
Astell Scientific™ Ethernet Interface
Export or print sterilization reports via this integrated ethernet port. Astell Scientific™ Ethernet Interface can also be used for remote monitoring, remote maintenance, and software updates.
Brand:  Astell Scientific™ AAR122
Product Code. 13417413
  • £332.00 / Each
Estimated Shipment: 04-11-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Enables secure network connectivity, remote access, and diagnostics.
  • Directly connects your autoclave to your own network.
  • Provides a fully secure connection between machine and Astell Scientific™, for the benefit of software updates, data interrogation and end-user troubleshooting or fault finding.
  • Configuration is a one-time affair at point of order.
  • Additional simple setup is easily completed by the end user.
  • Simplifies workflow and process and enables faster support to minimize possible down time.
Specifications
Specifications
Ethernet Interface
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Astell Scientific™ Ethernet Interface > Product Type: Ethernet Interface