missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss., absolute, sales not in Germany, ≥99.8% (GC), for license holders only, Honeywell Riedel-de Haën™
25.46 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Use promo code "21640" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
Stainless Steel Can
Specifications
Melting Point | -114°C (literature) |
CAS Max % | 100.0000% |
Density | 0.789 g/mL at 25 °C(lit.) |
Boiling Point | 78°C to 79°C |
Quantity | 1 L |
Flash Point | 12°C |
Packaging | Poly Bottle |
Linear Formula | CH3CH2OH |
UN Number | UN1170 |
Beilstein | 1718733 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'