Learn More
Invitrogen™ ESRRG Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
ESRRG (estrogen-related receptor gamma) ligand binding domain is a 54.05 kDa recombinant human protein (amino acids 229-458) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4417
Code : 50
Additional Details : Weight : 0.50000kg
Description
ERR γ-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Estrogen Related Receptor Gamma | |
Recombinant | |
Hormone Receptors |
2104 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Recombinant, Ligand Binding Domain | |
Human | |
GST Tag |