Learn More
Invitrogen™ ESRRB Human Protein, Ligand Binding Domain
Estrogen Related Receptor beta, Ligand Binding Domain (ERR beta-LBD) is a 54.29 kDa recombinant human protein (amino acids 204-433) expressed as a GST fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4802
Code : 50
Additional Details : Weight : 0.10500kg
Description
ERR beta was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Estrogen Related Receptor Beta, Ligand | |
Recombinant |
2103 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Ligand Binding Domain | |
Human | |
Hormone Receptors |