Learn More
ESRP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561779
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EFHMQHPSTC PARDLTVATM AQGLGLETDA TEDDFGVWEV KTMVAVILHL LKEPSSQLFS KPEVIKQKYE TGPCSKADVV DSETVV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
RBM35A, also known as ESRP2, is a mRNA splicing factor that with its related protein RBM35A (ESRP1) are coordinators of an epithelial cell-type-specific splicing program. Both RBM35B and RBM35A are involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs, suggesting that these proteins are global regulators of an epithelial regulatory network. Loss of this global ESRP-regulated epithelial splicing program induces the phenotypic changes in cell morphology that are observed during the epithelial-mesenchymal transition.Specifications
ESRP2 | |
Polyclonal | |
Unconjugated | |
ESRP2 | |
9530027K23Rik; Epithelial splicing regulatory protein 2; ESRP2; PP7059; RBM35B; RGD1310855; RNA binding motif protein 35A; RNA binding motif protein 35B; RNA-binding motif protein 35B; RNA-binding protein 35B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6T0 | |
ESRP2 | |
Recombinant protein corresponding to Human ESRP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only