Learn More
ERH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551723
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LVQPTKRPEG RTYADYESVN ECMEGVCKMY EEHLKRMNPN SPSITYDISQ LFDFIDDLAD LSCLVYRADT QTYQPYNKDW IKEKIYVLLR RQAQQAGK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
May have a role in the cell cycle. [UniProt]Specifications
ERH | |
Polyclonal | |
Unconjugated | |
ERH | |
DROER; Enhancer of rudimentary homolog; enhancer of rudimentary homolog (Drosophila); Erh; Mer; Prei1; preimplantation protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13877, 2079, 681415 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P84089, P84090 | |
ERH | |
Recombinant protein corresponding to Human ERH | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only