missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER Alpha Recombinant Protein, Ligand Binding Domain, GST Tag Recombinant Protein
ER Alpha Recombinant Protein, Ligand Binding Domain, GST Tag Recombinant Protein
Brand: Invitrogen A15677
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Estrogen Receptor (ER) Alpha Ligand Binding Domain (LBD) is a 62.80 kDa recombinant human protein (amino acids 282-595) expressed as a GST fusion protein in baculovirus-infected insect cells. It was expressed and purified in the absence of exogenous ligands. ER Alpha-LBD recombinant protein binds [3H]-estradiol with sub-nM affinity in hydroxylapatite assays and binds specifically to estrogen response element (ERE) sequences.ER Alpha-LBD recombinant protein is:
ER Alpha-LBD recombinant protein is manufactured under the strictest quality parameters. This product is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
Used in:
LanthaScreen™ TR-FRET ER Alpha Coactivator Assay Kit
LanthaScreen™ TR-FRET ER Alpha Competitive Binding Kit
For bulk reagents, email: !Mad-DiscoveryServices@lifetech.com
Specifications
NP_000116.2 | |
25 μg | |
Human | |
GST Tag | |
Insect | |
Recombinant, Ligand Binding Domain | |
Signal Transduction |
Nuclear Receptor Assay | |
estrogen receptor 1 (alpha); oestrogen receptor; NR3A1 | |
Recombinant | |
Suspended in: 50 mM Tris-HCl (pH 8.0), 500 mM KCl, 0.5 mM EDTA, 0.02% NP–40, 5 mM DTT, and 20% glycerol. | |
Growth Factors & Receptors | |
Liquid | |
Life Technologies™ |
For Research Use Only. Not for use in diagnostic procedures.