Learn More
Epstein Barr Virus Bcl-2 Monoclonal Antibody (D44E), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17275
Code : LD
Additional Details : Weight : 0.01000kg
Description
The MA1-7275 antibody reacts with Epstein-Barr virus Bcl-2 in viral and infected tissue samples. MA1-7275 has been successfully used in ELISA, Western blot, Immunohistochemical and Immunoprecipitation applications. The MA1-7275 antibody was raised against EBV BCl-2 homolog protein.
Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.Specifications
Epstein Barr Virus Bcl-2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
D44E | |
Unconjugated | |
EBV; EBV bcl2; EBV Bcl-2 | |
EBV BCl-2 homolog. | |
100 μg | |
Primary | |
4° C | |
Liquid |