Learn More
EPRS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555320
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CVQVVIIPCG ITNALSEEDK EALIAKCNDY RRRLLSVNIR VRADLRDNYS PGWKFNHWEL KGVPIRLEVG PRDMKSCQFV AVRRDTGEKL TVAENEAETK LQAIL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. EPRS is a multifunctional aminoacyl-tRNA synthetase that catalyzes the aminoacylation of glutamic acid and proline tRNA species.Specifications
EPRS | |
Polyclonal | |
Unconjugated | |
EPRS1 | |
2410081F06Rik; 3010002K18Rik; bifunctional aminoacyl-tRNA synthetase; Bifunctional glutamate/proline--tRNA ligase; C79379; Cell proliferation-inducing gene 32 protein; EARS; Eprs; EPRS1; GLNS; GLUPRORS; GluRS; glutamate tRNA ligase; Glutamate--tRNA ligase; Glutamatyl-prolyl-tRNA synthetase; glutamine-proline-tRNA synthetase; glutaminyl-tRNA synthetase; glutamyl-prolyl-tRNA synthetase; glutamyl-prolyl-tRNA synthetase 1; Glutamyl-tRNA synthetase; PARS; PIG32; proliferation-inducing gene 32 protein; proliferation-inducing protein 32; proline tRNA ligase; proline-tRNA ligase; Proline--tRNA ligase; prolyl-tRNA synthetase; ProRS; QARS; QPRS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P07814 | |
EPRS1 | |
Recombinant protein corresponding to Human EPRS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only