Learn More
Epiregulin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA546969
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Epiregulin is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Epiregulin Polyclonal antibody specifically detects Epiregulin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Neutralization, Western Blot.Specifications
Epiregulin | |
Polyclonal | |
Unconjugated | |
Ereg | |
Ep; Epiregulin; EPR; ER; EREG; Proepiregulin | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2069 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O14944 | |
Ereg | |
E. coli-derived recombinant human Epiregulin Val63-Leu108 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |