missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin B1/B3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538067
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Eph receptors and ephrins exhibit complementary expression in many tissues during embryogenesis indicating that bidirectional activation of Eph receptors and ephrin-B proteins may occur at expression domain interfaces. Ephrin-B1 transduces outside-in signals through C-terminal protein interactions that effect integrin-mediated cell attachment and migration. The distribution of ephrin-B1 in the developing retina suggests that it influences retinal axon mapping along the dorsal-ventral axis and may be involved in intratectal development The transmembrane ligand ephrin-B2 and its receptor tyrosine kinase EphB4 are specifically expressed on arterial and venous endothelial cells, respectively.Specifications
Ephrin B1/B3 | |
Polyclonal | |
Unconjugated | |
EFNB3 | |
CEK5 ligand, CEK5 receptor ligand, CEK5-L, CEL5-L, EFL-3, EFNB1, ELK ligand, ELK-L, EPH-related receptor tyrosine kinase ligand 2, EPL2, EPLG2, Ephrin-B1 precursor, LERK-2, LERK2, STRA1, STRA1 protein, kinase ephrin-B1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13641, 13643, 1947, 1949, 25186, 360546 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P98172, Q15768, O35393, P52795, P52796 | |
EFNB1, EFNB3 | |
A synthetic non-phosphopeptide derived from human Ephrin B around the phosphorylation site of Tyr330 (N-I-Y-Y-K) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |