Learn More
Ephrin A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545699
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EDSVPEDKTE RYVLYMVNFD GYSACDHTSK GFKRWECNRP HSPNGPLKFS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 86%
Ephrin A5 Polyclonal antibody specifically detects Ephrin A5 in Human samples. It is validated for Western Blot.Specifications
Ephrin A5 | |
Polyclonal | |
Unconjugated | |
EFNA5 | |
AF1; AL-1; AV158822; EFL5; EFL-5; EFNA5; EPH-related receptor tyrosine kinase ligand 7; ephrin A5; Ephrin-A5; Epl7; EPLG7; GLC1M; LERK7; LERK-7; RAGS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1946 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P52803 | |
EFNA5 | |
Synthetic peptide directed towards the N-terminal of human EFNA5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |