missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHB4 Recombinant Human Protein
EPHB4 Recombinant Human Protein
Brand: Invitrogen PV3251
Code : 50
Additional Details : Weight : 3.36000kg
Description
EPHB4 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation.Specifications
NP_004435 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
50.5kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
EPHB4, TYRO11 | |
EPHB4 was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 10% glycerol, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 561-987 | |
400nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Kinase and inhibitors | |
95% by SDS-PAGE |
0.35mg/mL | |
2050 | |
EPHB4 Recombinant Human Protein | |
–80°C | |
1 Each | |
EPHB4 Recombinant Human Protein | |
Ephrin and EPH receptor | |
EPHB4 Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.