Learn More
EphB3 Monoclonal Antibody (4A122D1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515394
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15394 targets EphB3 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15394 immunogen is purified recombinant fragment of EphB3 (aa39-212) expressed in E. Coli. MA5-15394 has been successfully used in immunohistochemistry analysis of Ephb3 in mouse E16.5 frozen lung sections.
EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.Specifications
EphB3 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P54753, P54754 | |
EPHB3 | |
Purified recombinant fragment of EphB3 (aa39-212) expressed in E. Coli. | |
RUO | |
13845, 2049 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4A122D1 | |
Unconjugated | |
EPHB3 | |
AW456895; Cek10; Developmental kinase 5; EK2; Embryonic kinase 2; EPH receptor B3; EPHB3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin type-B receptor 3; Etk2; HEK2; human embryo kinase 2; Mdk5; mDK-5; Sek4; TYRO6; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |