missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB Pan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539736
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.Specifications
EphB Pan | |
Polyclonal | |
Unconjugated | |
EPHB3 | |
ETK2; HEK2; TYRO6; EPHB3, DRT; ERK; CAPB; Hek5; PCBC; EPHT3; Tyro5; MGC87492; EPHB2, Ephrin type-B receptor 1; Tyrosine-protein kinase receptor EPH-2; NET, HEK-6, ELK, Ephrin type-B receptor 2, Ephrin type-B receptor 3, Ephrin B1, EphrinB1, EphrinB3, Ephrin B3, Ephrin B2, EphrinB2, Ephrin B4, EphrinB4, Ephrin type-B receptor 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13844, 13845, 13846, 2047, 2048, 2049, 2050, 270190 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P29323, P54753, P54760, P54762, P54754, P54761, P54763, Q8CBF3 | |
EPHB1, EPHB2, EPHB3, EPHB4 | |
A synthetic non-phosphopeptide derived from human EPHB1/2/3/4 around the phosphorylation site of Tyr600/602/614/596 (F-T-YP-E-D) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |