missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 347400
Code : 50
Additional Details : Weight : 0.50000kg
Description
EphA2 Polyclonal antibody specifically detects EphA2 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
EphA2 | |
Polyclonal | |
Unconjugated | |
EPHA2 | |
ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1969 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P29317 | |
EPHA2 | |
Synthetic peptide derived from the C-terminal region of the human EphA2 receptor protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |