missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM Monoclonal Antibody (VU-1D9), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15755
Code : NEW
Additional Details : Weight : 0.00280kg
Description
EGP40 is a 40kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.Specifications
EpCAM | |
Monoclonal | |
FITC | |
EPCAM | |
Epithelial Specific Antigen, ESA, KSA, M4S1, MK-1, DIAR5, EGP-2, EGP40, KS1/4, MIC18, TROP1, TACSTD1, EPCAM, Epithelial cell adhesion molecule | |
Mouse | |
Purified | |
RUO | |
4072 | |
4° C | |
Liquid |
Flow Cytometry | |
VU-1D9 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P16422 | |
EPCAM | |
Human CD326 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |