missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525141
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Xenopus based on sequence homology.
EPC2 may play a role in transcription or DNA repair.Specifications
EPC2 | |
Polyclonal | |
Unconjugated | |
EPC2 | |
5830499L14; D2Ertd694e; Enhancer of polycomb homolog 2; enhancer of polycomb homolog 2 (Drosophila); Epc2; EPC-LIKE | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
227867, 26122 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q52LR7, Q8C0I4 | |
EPC2 | |
KLH conjugated synthetic peptide between 205-233 amino acids from the N-terminal region of human EPC2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |