Learn More
EPC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540701
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: VNKQDKADLI RPKRKYEKKP KVLPSSAAAT PQQTSPAALP VFNAKDLNQY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
EPC1 Polyclonal antibody specifically detects EPC1 in Human samples. It is validated for Western Blot.Specifications
EPC1 | |
Polyclonal | |
Unconjugated | |
EPC1 | |
2400007E14Rik; 5730566F07Rik; A930032N02Rik; DKFZp781P2312; enhancer of polycomb 1; enhancer of polycomb homolog 1; enhancer of polycomb homolog 1 (Drosophila); Epc1; Epl 1; Epl1; mir-1893 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
80314 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H2F5 | |
EPC1 | |
Synthetic peptide directed towards the middle region of human EPC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |