missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41L5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549177
Additional Details : Weight : 0.01000kg
Description
Predicted to react with mouse based on sequence homology.
May contribute to the correct positioning of tight junctions during the establishment of polarity in epithelial cells.Specifications
EPB41L5 | |
Polyclonal | |
Unconjugated | |
EPB41L5 | |
Band 4.1-like protein 5; BE37; EPB41L5; erythrocyte membrane protein band 4.1 like 5; KIAA1548; YMO1; YRT; yurt homolog | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
57669 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9HCM4 | |
EPB41L5 | |
KLH conjugated synthetic peptide between 355-383 amino acids from the Central region of human EPB41L5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |