Learn More
EPB41L3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518453
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
Critical growth regulator in the pathogenesis of meningiomas. Its human homolog acts as a tumor suppressor and binds 14-3-3 proteins.Specifications
EPB41L3 | |
Polyclonal | |
Unconjugated | |
EPB41L3 | |
130kDa Protein 4.1B MEF cell isoform; 4.1B; 60kDa 4.1B MEF cell isoform; band 4.1-like protein 3; Band 4.1-like protein 3, N-terminally processed; Dal1; DAL-1; DAL1P; differentially expressed in adenocarcinoma of the lung protein 1; Epb4.1l3; Epb41l3; epb41l3.L; erythrocyte membrane protein band 4.1 like 3; erythrocyte membrane protein band 4.1-like 3; erythrocyte membrane protein band 4.1-like 3 L homeolog; erythrocyte protein band 4.1 like 3; erythrocyte protein band 4.1-like 3; FLJ37633; HGNC:3380; Kiaa0987; mDAL-1; NBL3; XELAEV_18031863mg | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23136 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y2J2 | |
EPB41L3 | |
Synthetic peptide sequence (HKETEITPEDGED) corresponding to the C-terminus amino acids of EPB41L3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |