Learn More
Merck ZHE Hazardous Waste System
ZHE (Zero Head Space Extractor) system has been designated by the USEPA as a suitable apparatus for Toxicity Characteristic Leaching Procedure (TCLP).
Brand: Merck YT30090HW
4065.25 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 72
Additional Details : Weight : 0.10000kg
Description
Cellulose support pads are used to reinforce filters in Millipore's monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.To increase flow rate and throughput, our woven mesh spacers can be placed between filters in a serial filtration to prevent the downstream screen filter from in.blinding in. the upstream filter pores.
Features & Benefits
- Avoid membrane damage during high pressure or fast flow conditions
- Spacers enable you to combine multiple stages of filtration into a single process step, saving time
Applications: Air Monitoring, Environmental Monitoring, Aerosol Contamination Monitoring
Compliance
Designated by the U.S. EPA as a suitable apparatus for TCLP
Specifications
Not Autoclavable | |
316 Stainless Steel Body; Molded PP Handwheels; Fluoroelastomer-A O-Rings | |
3.5 bar | |
Environmental Test Kits or Supplies | |
15.9 cm |
Hazardous Waste System | |
Non-sterile | |
3.5 bar | |
27.9 cm |