missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck OptiScale™ Capsule Filters for Aseptic Processing Applications
Small-volume option for process screening and scaling
£505.00
Specifications
Sterility | Non-sterile |
---|---|
Filtration Area | 17.7 cm2 |
For Use With (Application) | Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates |
Housing Material | Polycarbonate |
Product Code | Brand | Pore Size | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
15926255
|
Merck
SHGEA47HH3 |
0.5/0.2 μm | 3.7 in. | 9.4 cm |
£505.00
Each |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- With “drop-in” filters
- Non-sterile
- PVDF vent cap
- Wettability: Hydrophilic
- Housing and drainage disk: Polycarbonate
- Filter support: Polypropylene
- Filtration Area: 17.7cm2
Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates
Specifications
Non-sterile | |
17.7 cm2 | |
Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates | |
Polycarbonate |