Learn More
Merck MultiScreen™HTS Vacuum Manifold
For vacuum filtration using 96- and 384-well MultiScreenHTS, MultiScreen™ Solvinert, and Solvinert Deep Well filter plates
Brand:  Merck MSVMHTS00
Code : 73
Additional Details : Weight : 5.44310kg
Product Code. 10106982
  • £2850.00 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Includes: Manifold base, standard collar, gaskets and gasket inserts, tubing, valves, pressure gauge
Description
Description
  • Accepts deep well or standard receiver plates Suitable for use with manual or automated protocols; ANSI/SBS compliant footprint allows for easy robotic deck integration
  • Easily configured for both flow-to-waste and analyte collection
  • DirectStack™ technology for crosstalk-free filtrate collection
  • Manifold components are resistant to solvents
Specifications
Specifications
MultiScreen Vacuum Manifold
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products