missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™HTS 96-Well Filter Plates for Clearing
Designed to meet specific performance criteria for sample preparation methodology
Marca: Merck MSNANLY50
Code : NEW
Detalles adicionales : Weight : 0.10000kg
Descripción
- Automation-compatible multiscreen filter plates
- Lysate clearing
Clearing of bacterial cell lysates
Especificaciones
Standard | |
8 x 12 | |
Clear | |
96 | |
0.26 cm2 | |
50 | |
1.28 cm | |
1.44 cm |
Negatively Charged Phosphocellulose, Polyester Mesh | |
0.65 μm | |
Barex | |
Non-sterile | |
Protein kinase assays with peptide substrates, enzyme assays, binds positive charge | |
0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44 cm) | |
8.55 cm |