missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millistak+™ HC Pod Disposable Depth Filter System
Combine two distinct technologies to enhance filter capacity and retention
£357.00
Specifications
Disposable | Single-use |
---|---|
Material | Glass Fiber-reinforced Polypropylene |
Material (Membrane) | Cellulose |
Inlet Connection (English) | 0.25 in. hose |
Max. Pressure | 50 psi at 25°C, 15 psi at 80°C |
Product Code | Brand | Grade | Filtration Area | For Use With (Application) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Grade | Filtration Area | For Use With (Application) | Price | Quantity & Availability | |||
15822305
|
Merck
MC0HC027H1 |
C0HC | 0.027 m2 | Perfusion bioreactor fluid, laboratory filtration |
£357.00
1 set |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Beschreibung
- Filter material: cellulose and inorganic filter aid
- Configurations: 0.11, 0.55, or 1.1 m2 filtration area
- Low hold-up volume for greater product yield
- Broad range of 23 media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000L or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Disposable Pod system protects operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Ideal for Primary and Secondary Clarification in Lab, Pilot and Process-Scale Applications
Spezifikation
Single-use | |
Cellulose | |
50 psi at 25°C, 15 psi at 80°C | |
Millistak+ Lab Scale Pod Filter | |
22 cm | |
14 cm |
Glass Fiber-reinforced Polypropylene | |
0.25 in. hose | |
1 Ea. | |
8.5 in. | |
5.3 in. |