Learn More
Merck Millipore Millipak™-20 Filter Unit
Non-sterile filter removes 0.22μm particles at point-of-use prior to dispensing
Brand:  Merck Millipore MPGP02001
Code : NEW
Additional Details : Weight : 0.10800kg
Product Code. 10740365
  • £322.00 / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
Non-sterile filter removes 0.22μm particles at point-of-use prior to dispensing
  • Standard point-of-use filter
  • Nonsterile