missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Milli-Q™ Advantage A10 Ultrapure Water Purification System Accessory, Quantum Polishing Cartridges
Quantum polishing catridge removes ionic and organic contaminants below trace levels to match required water quality requirements
£835.00
Specifications
For Use With (Equipment) | Milli-Q™ Systems |
---|---|
For Use With (Application) | Spectrophotometry, spectroscopy, chromatography, and biochemistry |
Product Code | Brand | Type | Resin Type | Purification Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Resin Type | Purification Method | Price | Quantity & Availability | |||
10176094
|
Merck Millipore
QTUM0TEX1 |
Quantum TEX Cartridge Filter | Organex & Ionex | Ion Exchange, Activated Carbon |
£835.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||
Description
Quantum polishing catridge removes ionic and organic contaminants below trace levels to match required water quality requirements
Quantum™ TEX Features
- Contains Ionex resins for trace ion levels
- Feedwater nature: Ultrapure
- Technology: Activated carbon, Ion exchange
- Contains Organex and Ionex resins for trace levels of organics and ions
- Feedwater nature: Ultrapure
- Technology: Activated carbon, Ion exchange
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Certifications
ISO 9001 v.2000 and ISO 14001
Compliance
USP 645 suitability requirements
Specifications
Milli-Q™ Systems | |
Spectrophotometry, spectroscopy, chromatography, and biochemistry |