Learn More
Greiner Bio-One Easyload Pipette Tip Refill System, Sterile
Stacked refill system with rack suitable for refilling pipette tip boxes.
Specifications
Sterility | Sterile |
---|---|
Autoclavable | Autoclavable |
Sterilization Method | Irradiation |
Disposable | Single-use |
Graduated | Yes |
Description
- Simple, time- and space-saving refill system for standard pipette tips.
- Autoclavable racks.
- Stackable racks.
- Color-coded boxes.
- Positioning guides make it easier to place refill units on the racks.
- Design of rack interior increases stability during autoclaving and when pipette tips are bein used with multi-channel pipettes.
- Graduated
- Sterile
- For single use only
- Polypropylene construction
- Free of heavy metal
Specifications
Sterile | |
Irradiation | |
Yes | |
Polypropylene |
Autoclavable | |
Single-use | |
EasyLoad |