missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ 96 Agarose Gels with SYBR™ Safe DNA Gel Stain, 2%

Designed for fast, convenient, and safe DNA sample electrophoresis

£210.00

Specifications

Compatible Ladders E-Gel™ Low Range Quantitative DNA Ladder
Gel Percentage 2%
Gel Type E-Gel
Product Type Agarose Gel
Separation Range 100 to 2000 bp
View More Specs

Products 1
Product Code Brand Price Quantity & Availability  
Product Code Brand Price Quantity & Availability  
10063143
View Documents Promotion Details
Invitrogen™
G720802
8 gels
£210.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
 
Description

Description

  • Bufferless, precast gels designed for fast, high-throughput DNA electrophoresis
  • Each gel contains 96 sample lanes and 8 marker lanes that for screening multiple PCR products, plasmid preparations, and restriction digests
  • The staggered-well format provides a 1.6cm run length that resolves between 100 bp and 10 kb
  • Loading format compatible with multi-channel pipettors, and the most commonly used 8-, 12-, and 96-pin liquid handling robots
  • With 12-minute run times, up to 20,000 samples can be resolved in a single day
  • Unlike ethidium bromide, SYBR Safe DNA Gel Stain is a nontoxic, nonmutagenic DNA stain that is safer for the user and for the environment

Applications:

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Specifications

Specifications

E-Gel™ Low Range Quantitative DNA Ladder
E-Gel
100 to 2000 bp
Less hazardous
2%
Agarose Gel
SYBR™ Safe
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.