missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Dystrophin, Rabbit Polyclonal Antibody.
£86.09 - £480.42
Specifications
Antigen | Dystrophin |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12683337
|
Epredia
RB-9024-P1 |
Concentrated | 500 μL |
£390.00
500µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12613347
|
Epredia
RB-9024-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12603347
|
Epredia
RB-9024-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12623347
|
Epredia
RB-9024-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immuohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide corresponding to C-terminal of human Dystrophin
Molecular Weight: 427 kDa
Positive Control: Skeletal muscle
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Dystrophin | |
Polyclonal | |
Rabbit | |
Muscle Biology | |
A synthetic peptide corresponding to C-terminal of human Dystrophin | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG |