Learn More
DyNAmo Flash DyNAmo Flash Probe qPCR Kit
Perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput.
£129.00 - £1992.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Tbr DNA polymerase |
Description
DyNAmo Flash Probe qPCR Kit is the perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real-time PCR instruments. Performance of the DyNAmo Flash Probe qPCR Kit is based on an efficient hot start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results.
DyNAmo Flash Probe qPCR Kit is optimized for hydrolysis probes (e.g., TaqMan probes) and it can be used with both block- and capillary-based realtime PCR instruments.
- Extremely fast protocols: combined annealing and extension step of only 15 sec.
- Specific and sensitive detection of a wide range of template concentrations
- Included dUTP allows the use of UNG for prevention of carry-over contamination
- Convenient 2X master mix and optimized protocols
Specifications
Primer-probe | |
High | |
Tbr DNA polymerase | |
DyNAmo Flash Probe qPCR Kit | |
Real Time PCR SYBR Master Mix | |
2X (DyNAmo Flash qPCR Mix), 50X (ROX Dye) |
Tube | |
qPCR | |
Fast | |
DyNAmo Flash | |
Gene Expression | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.