missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Amber, USP /EP (3.2.1) Light Transmission

Catalog Number p-7189456
569.99 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.


Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
50 mL
500 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Classic wide-neck, glass-stoppered reagent bottles in ambered glass

The classical stoppered reagent bottles are moulded from amber DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel. The amber glass ensures that light sensitive contents are protected from the effects of photo-degradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle.

  • USP/EP UV protection up to approx. 500 nm due to external colouration
  • The amber colour is only applied externally ensuring that internal surfaces have standard DURAN™ clear Type 1 glass properties
  • Very durable and chemically resistant amber colour
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • All glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
Capacity (Metric) 250 mL
Description Neck with standard ground joint and glass flat-headed stopper
For Use With (Application) Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Autoclavable Autoclavable
Diameter (Metric) 70 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'