missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
Description
Open topped cap with central aperture suitable for use with bromobutyl stoppers, silicone piercing septa or glassware connections. More chemically resistant than PP cap.
- Compatible with GL bottle threads
- Red PBT offers high temperature and chemical resistance
- Glass reinforced PBT offers high strength
Specifications
Specifications
Color | Red |
Diameter (Metric) Inner | Bore Size 11 mm |
For Use With (Application) | Aseptic reagent delivery and culture sampling using a syringe and needle |
Material | PBT |
Quantity | 10/Pk. |
Screw Material | Polybutylene Terephthalate (PBT) with glass fiber |
Thread Size (Metric) | GL 18 |
Autoclavable | Autoclavable |
Product Type | Open Top Screw Cap |
Closure Material | PBT with Glass Fiber |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'